ENA432540428 Enamine

2-[(prop-2-yn-1-yl)amino]acetonitrile

  • MDL number MFCD00137987
  • Cas Number : 56096-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-199311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199311
Availability details for
Quantity
  • Availability
  • Add to Cart