ENA432540433 Enamine

3-carbamoyl-5-chlorobenzene-1-sulfonyl chloride

  • MDL number MFCD21196017
  • Cas Number : 1490401-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199316
Availability details for
Quantity
  • Availability
  • Add to Cart