ENA432540436 Enamine

5-{[(tert-butoxy)carbonyl]amino}thiophene-2-carboxylic acid

  • MDL number MFCD12177582
  • Cas Number : 1094071-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199331
Availability details for
Quantity
  • Availability
  • Add to Cart