ENA432540437 Enamine

3-ethyl-4-hydroxybenzonitrile

  • MDL number MFCD16997588
  • Cas Number : 4997-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198649
Availability details for
Quantity
  • Availability
  • Add to Cart