ENA432540441 Enamine

2-[(carboxymethyl)[(pyridin-3-yl)methyl]amino]acetic acid hydrochloride

  • MDL number MFCD28145632
  • Cas Number : 1803611-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199349
Availability details for
Quantity
  • Availability
  • Add to Cart