ENA432540442 Enamine

1,7,7-trimethylspiro[bicyclo[2.2.1]heptane-2,4'-imidazolidine]-2',5'-dione

  • MDL number MFCD13818260
  • Cas Number : 6929-30-2
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198650
Availability details for
Quantity
  • Availability
  • Add to Cart