ENA432540445 Enamine

4-(2-hydroxypropanoyl)benzonitrile

  • MDL number MFCD24697625
  • Cas Number : 1266343-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-198652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198652
Availability details for
Quantity
  • Availability
  • Add to Cart