ENA432540469 Enamine

N-(1-cyano-1-methylethyl)acetamide

  • MDL number MFCD01733809
  • Cas Number : 40652-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25603
Availability details for
Quantity
  • Availability
  • Add to Cart