ENA432540470 Enamine

1-(2-methyl-5-nitrobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD28145642
  • Cas Number : 1803607-05-7
  • Molecular Weight : 321.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-((2-methyl-5-nitrophenyl)sulfonyl)piperazine hydrochloride
Inchi Code 1S/C11H15N3O4S.ClH/c1-9-2-3-10(14(15)16)8-11(9)19(17,18)13-6-4-12-5-7-13;/h2-3,8,12H,4-7H2,1H3;1H
Inchi Key KAOMPYHOBALVGL-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-30113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30113
Availability details for
Quantity
  • Availability
  • Add to Cart