ENA432540479 Enamine

2-(piperazine-1-sulfonyl)benzonitrile hydrochloride

  • MDL number MFCD28145648
  • Cas Number : 1803586-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-30166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30166
Availability details for
Quantity
  • Availability
  • Add to Cart