ENA432540480 Enamine

1-(2-bromobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD08064298
  • Cas Number : 864759-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-30171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30171
Availability details for
Quantity
  • Availability
  • Add to Cart