ENA432540481 Enamine

(1-cyclopropylethyl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD28139644
  • Cas Number : 1803600-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30176
Availability details for
Quantity
  • Availability
  • Add to Cart