ENA432540484 Enamine

2-[(piperazin-1-yl)methyl]-1,3-benzothiazole dihydrochloride

  • MDL number MFCD10599641
  • Cas Number : 1171038-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30200
Availability details for
Quantity
  • Availability
  • Add to Cart