ENA432540488 Enamine

1-(2,5-difluorobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD28145650
  • Cas Number : 1568672-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-30291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30291
Availability details for
Quantity
  • Availability
  • Add to Cart