ENA432540490 Enamine

1-[(dimethyl-1,2-oxazol-4-yl)sulfonyl]piperazine hydrochloride

  • MDL number MFCD28145652
  • Cas Number : 1803583-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-100
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30304
Availability details for
Quantity
  • Availability
  • Add to Cart