ENA432540493 Enamine

N-(5-methyl-1,2-oxazol-3-yl)-2-(piperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD10597397
  • Cas Number : 1172414-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30578
Availability details for
Quantity
  • Availability
  • Add to Cart