ENA432540494 Enamine

1-[(2,3-dihydro-1-benzofuran-5-yl)methyl]piperazine

  • MDL number MFCD07366361
  • Cas Number : 55745-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-231
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31934
Availability details for
Quantity
  • Availability
  • Add to Cart