ENA432540499 Enamine

1-(1-methyl-4-nitro-1H-imidazol-5-yl)piperazine hydrochloride

  • MDL number MFCD28145654
  • Cas Number : 1803597-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34529
Availability details for
Quantity
  • Availability
  • Add to Cart