ENA432540502 Enamine

1-[(4,5-dimethoxy-2-methylphenyl)methyl]piperazine

  • MDL number MFCD05189182
  • Cas Number : 523980-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34782
Availability details for
Quantity
  • Availability
  • Add to Cart