ENA432540503 Enamine

1-{[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine hydrochloride

  • MDL number MFCD22421961
  • Cas Number : 1417566-84-7
  • Molecular Weight : 294.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(piperazin-1-ylmethyl)-3-(p-tolyl)-1,2,4-oxadiazole hydrochloride
Inchi Code 1S/C14H18N4O.ClH/c1-11-2-4-12(5-3-11)14-16-13(19-17-14)10-18-8-6-15-7-9-18;/h2-5,15H,6-10H2,1H3;1H
Inchi Key XSEFGNFSNPTGQF-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-34835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34835
Availability details for
Quantity
  • Availability
  • Add to Cart