ENA432540504 Enamine

3-(2-fluorophenyl)-1-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD09749449
  • Cas Number : 1314962-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-198665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198665
Availability details for
Quantity
  • Availability
  • Add to Cart