ENA432540506 Enamine

2-[(piperazin-1-yl)methyl]-5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06655575
  • Cas Number : 900640-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35255
Availability details for
Quantity
  • Availability
  • Add to Cart