ENA432540511 Enamine

1H,3H,4H,9H-pyrano[3,4-b]indole-1,3-dione

  • MDL number MFCD03828555
  • Cas Number : 60442-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42654
Availability details for
Quantity
  • Availability
  • Add to Cart