ENA432540522 Enamine

6-chloro-9-ethyl-9H-purine

  • MDL number MFCD07440136
  • Cas Number : 5462-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63513
Availability details for
Quantity
  • Availability
  • Add to Cart