ENA432540526 Enamine

2-bromo-5-chloro-3-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD17240570
  • Cas Number : 1340462-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66818

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-66818
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart