ENA432540531 Enamine

4-chloro-8-propoxyquinoline

  • MDL number MFCD12192754
  • Cas Number : 1156602-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67703
Availability details for
Quantity
  • Availability
  • Add to Cart