ENA432540536 Enamine

7-chloro-3-methyl-1H-pyrazolo[4,3-d]pyrimidine

  • MDL number MFCD08695123
  • Cas Number : 5399-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-155
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72256
Availability details for
Quantity
  • Availability
  • Add to Cart