ENA432540539 Enamine

2-(cyclopropylmethoxy)pyridine-4-carbonitrile

  • MDL number MFCD11136691
  • Cas Number : 1019568-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73288
Availability details for
Quantity
  • Availability
  • Add to Cart