ENA432540550 Enamine

N-methyl-6-nitropyridin-3-amine

  • MDL number MFCD16663398
  • Cas Number : 189348-22-9
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl-6-nitro-3-pyridinamine
Inchi Code 1S/C6H7N3O2/c1-7-5-2-3-6(8-4-5)9(10)11/h2-4,7H,1H3
Inchi Key NAXPEXYGRNHRSQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85813

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-85813
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart