ENA432540554 Enamine

5-hydroxy-4,4-dimethylpentanenitrile

  • MDL number MFCD11499290
  • Cas Number : 25252-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90376
Availability details for
Quantity
  • Availability
  • Add to Cart