ENA432540556 Enamine

methyl 3-(trifluoromethyl)pyrrolidine-3-carboxylate

  • MDL number MFCD18870232
  • Cas Number : 1283851-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91067
Availability details for
Quantity
  • Availability
  • Add to Cart