ENA432540557 Enamine

2-(1,3-thiazol-5-yl)acetonitrile

  • MDL number MFCD12402326
  • Cas Number : 50382-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91125
Availability details for
Quantity
  • Availability
  • Add to Cart