ENA432540558 Enamine

3-amino-4-methylpiperidin-2-one

  • MDL number MFCD19208224
  • Cas Number : 53611-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91401
Availability details for
Quantity
  • Availability
  • Add to Cart