ENA432540561 Enamine

3-bromo-2-fluoro-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD21249810
  • Cas Number : 1485934-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92246
Availability details for
Quantity
  • Availability
  • Add to Cart