ENA432540563 Enamine

N-[(1-phenyl-1H-pyrazol-4-yl)methylidene]hydroxylamine

  • Cas Number : 23972-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93337
Availability details for
Quantity
  • Availability
  • Add to Cart