ENA432540564 Enamine

methyl 6-aminopyridazine-3-carboxylate

  • MDL number MFCD11226154
  • Cas Number : 98140-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-93549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93549
Availability details for
Quantity
  • Availability
  • Add to Cart