ENA432540575 Enamine

tert-butyl N-(3-oxopyrazolidin-4-yl)carbamate

  • MDL number MFCD13176501
  • Cas Number : 106693-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99847
Availability details for
Quantity
  • Availability
  • Add to Cart