ENA444722709 Enamine

tert-butyl 1-{[(1-methyl-1H-imidazol-2-yl)methyl]amino}-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD28348001
  • Cas Number : 1803583-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159224
Availability details for
Quantity
  • Availability
  • Add to Cart