ENA444722784 Enamine

1-(azetidin-3-yl)ethan-1-one; trifluoroacetic acid

  • MDL number MFCD28246274
  • Cas Number : 1309208-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-196174
Availability details for
Quantity
  • Availability
  • Add to Cart