ENA444722786 Enamine

1-amino-4-chloropentan-2-one hydrochloride

  • MDL number MFCD28246275
  • Cas Number : 1803584-26-0
  • Molecular Weight : 172.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196344
Availability details for
Quantity
  • Availability
  • Add to Cart