ENA444722808 Enamine

1-(4-benzylmorpholin-2-yl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD28246294
  • Cas Number : 1807891-11-7
  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197515
Availability details for
Quantity
  • Availability
  • Add to Cart