ENA444722810 Enamine

lithium(1+) ion (5S)-5-(4-fluorophenyl)-5-hydroxypentanoate

  • MDL number MFCD28246297
  • Cas Number : 1807901-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-197677
Availability details for
Quantity
  • Availability
  • Add to Cart