ENA444722826 Enamine

1-[(1-methyl-1H-pyrazol-4-yl)methyl]piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD28246320
  • Cas Number : 1803565-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-197981
Availability details for
Quantity
  • Availability
  • Add to Cart