ENA444722831 Enamine

methyl 4-phenoxypyridine-3-carboxylate

  • MDL number MFCD28246326
  • Cas Number : 845671-56-9
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198165
Availability details for
Quantity
  • Availability
  • Add to Cart