ENA444722848 Enamine

2-(3-fluorophenyl)pyrazine

  • MDL number MFCD28246349
  • Cas Number : 1286755-19-8
  • Molecular Weight : 174.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198391
Availability details for
Quantity
  • Availability
  • Add to Cart