ENA444722851 Enamine

4-(3-chlorophenyl)-4-fluoropiperidine hydrochloride

  • MDL number MFCD28246355
  • Cas Number : 1803581-95-4
  • Molecular Weight : 250.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-198437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198437
Availability details for
Quantity
  • Availability
  • Add to Cart