ENA444722877 Enamine

[1-(methylsulfanyl)cyclobutyl]methanol

  • MDL number MFCD28246384
  • Cas Number : 84657-77-2
  • Molecular Weight : 132.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199351
Availability details for
Quantity
  • Availability
  • Add to Cart