ENA444722883 Enamine

tert-butyl N-[3-(3-fluorophenyl)prop-2-yn-1-yl]carbamate

  • MDL number MFCD28348054
  • Cas Number : 1803599-03-2
  • Molecular Weight : 249.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199501
Availability details for
Quantity
  • Availability
  • Add to Cart