ENA444722947 Enamine

3-(1-methyl-1H-imidazol-2-yl)cyclohexan-1-one

  • MDL number MFCD20336516
  • Cas Number : 1343342-67-5
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-189239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189239
Availability details for
Quantity
  • Availability
  • Add to Cart