ENA444722992 Enamine

(1,2,4-benzotriazin-3-yl)methanol

  • MDL number MFCD28246244
  • Cas Number : 342410-94-0
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-192030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192030
Availability details for
Quantity
  • Availability
  • Add to Cart